Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 7
Recommended positive controls: 293T, A431, HeLa, HepG2.
Predicted reactivity: Mouse (91%), Rat (90%), Zebrafish (81%), Pig (91%), Chicken (88%), Chimpanzee (100%), Bovine (89%).
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
The 90 kDa heat shock protein (hsp90) family of molecular chaperones is a highly conserved family of proteins that play an important physiological role. Hsp90 is involved in numerous cellular processes but is best known for its association with signal transduction machinery. A recently cloned homolog of hsp90 is the tumor necrosis factor receptor-associated protein (TRAP1). Like hsp90, TRAP1 is found to be associated with numerous proteins involved in diverse actions. Immunofluorescence data has shown TRAP1 to be localized in the mitochondria of mammalian cells. This observation and the fact that TRAP1 is shown to have a mitochondrial targeting presequence strongly implicates TRAP1 as a mitochondrial matrix protein.
⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Heat shock protein 75 kDa, mitochondrial; Heat shock protein family C member 5; HSP 75; testicular tissue protein Li 209; TNF receptor-associated protein 1; TNFR-associated protein 1; TRAP-1; Tumor necrosis factor type 1 receptor-associated protein
Gene Aliases: 2410002K23Rik; HSP 75; HSP75; HSP90L; HSPC5; TRAP-1; TRAP1
UniProt ID: (Human) Q12931, (Mouse) Q9CQN1
Entrez Gene ID: (Human) 10131, (Mouse) 68015
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support