Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 2
CpG methylation is an epigenetic modification that is important for embryonic development, imprinting, and X-chromosome inactivation. Studies in mice have demonstrated that DNA methylation is required for mammalian development. This gene encodes a protein with similarity to DNA methyltransferases, but this protein does not display methyltransferase activity. The protein strongly binds DNA, suggesting that it may mark specific sequences in the genome. Alternative splicing results in multiple transcript variants encoding different isoforms.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: DNA (cytosine-5)-methyltransferase-like protein 2; DNA cytosine-5 methyltransferase 2; DNA methyltransferase 2; DNA methyltransferase homolog HsaIIP; DNA methyltransferase homolog MmuIIP; DNA methyltransferase-2; DNA MTase homolog HsaIIP; DNA MTase homolog MmuIIP; Dnmt2; M.HsaIIP; m.MmuIIP; Met-2; PuMet; RP11-406H21.1; tRNA (cytosine(38)-C(5))-methyltransferase; tRNA (cytosine-5-)-methyltransferase
Gene Aliases: DMNT2; DNMT2; Met2; MHSAIIP; PUMET; RNMT1; Rnmt2; TRDMT1
UniProt ID: (Human) O14717, (Mouse) O55055
Entrez Gene ID: (Human) 1787, (Mouse) 13434
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support