Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Percent identity with other species by BLAST analysis: Human, Gorilla, Gibbon, Monkey, Sheep, Dog, Elephant, Panda, Horse, Pig (100%); Marmoset, Mouse, Rat, Bovine, Hamster, Opossum, Turkey, Chicken, Lizard, Xenopus (94%); Stickleback (88%); Rabbit, Zebrafish (81%).
For IHC(P), use heat induced antigen retrieval in pH 6.0 citrate buffer. After incubation with the primary antibody, slides were incubated with biotinylated secondary antibody, followed by alkaline phosphatase-streptavidin and chromogen.
TRH-R is a 398 amino acid integral membrane protein with six transmembrane domains and belongs to G-protein coupled receptor 1 family. It acts as a receptor for the signaling hormone TRH and activates the inositol phospholipid-calcium-protein kinase C transduction pathway upon binding with TRH. Reports suggest that TRH-R is a neurotransmitter and neuro regulator that controls dimerization of the TRH receptor and potentiates hormone-induced receptor trafficking. It is known to widely express in the thyrotroph cells of the anterior pituitary.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: MGC141920; Thyroliberin receptor; Thyrotropin-releasing hormone receptor; TRH receptor; TRH-R; TRH1 Receptor
Gene Aliases: TRH-R; TRH-RECEPTOR; TRHR
UniProt ID: (Human) P34981, (Sheep) Q28596
Entrez Gene ID: (Pig) 100415777, (Dog) 482007, (Human) 7201, (Horse) 100056618, (Sheep) 443425
Molecular Function:
C4 zinc finger nuclear receptor
DNA-binding transcription factor
zinc finger transcription factor
gene-specific transcriptional regulator
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support