Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Cell lysate).
Immunogen sequence: PDRATAVATA SRLGPYVLLE PEEGGRAYRA LHCPTGTEYT CKVYPVQEAL AVLEPYARLP PHKHVARPTE VLAGTQLLYA FFTRTHGDMH
A novel inhibitor of cell division, Tribbles, blocks mitosis at a critical point in Drosophila morphogenesis. Recently, mammalian homologues of Tribbles (Trbs) have been identified that control mitogen-activated protein kinase (MAPK) activity. TRIB3 also known as TRB-3, SINK, and SKIP3 is a member of Trbs family. Overexpression of this protein inhibits NF-kB-dependent transcription induced by tumor necrosis factor (TNF) stimulation or its downstream signaling proteins but does not inhibit NF-kB translocation to the nucleus and binding to DNA. TRIB3/SKIP3 is also overexpressed in multiple human tumors and is regulated by hypoxia.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: DJ1103G7.3; neuronal cell death inducible putative kinase; Neuronal cell death-inducible putative kinase; p65-interacting inhibitor of NF-kappa-B; p65-interacting inhibitor of NF-kappaB; SINK; TRB-3; Tribbles 3; Tribbles homolog 3
Gene Aliases: C20orf97; NIPK; SINK; SKIP3; TRB3; TRIB3
UniProt ID: (Human) Q96RU7
Entrez Gene ID: (Human) 57761
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support