Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MPATPSLKV VHELPACTLC AGPLEDAVTI PCGHTFCRLC LPALSQMGAQ SSGKILLCPL CQEEEQAETP MAPVPLGPLG ETYCEEHGEK IYFFCENDAE FLCVFCREGP THQAHTVGFL DEAIQPYRDR LRSRLEALST ERDEIEDVKC QEDQKLQVLL TQIESKKHQV ETAFERLQQE LEQQRCLLLA RLRELEQQIW KERDEYITKV SEEVTRLGAQ VKELEEKCQQ PASELLQDVR VNQSRCEMKT FVSPEAISPD LVKKIRDFHR K (1-270 aa encoded by BC038585)
The protein encoded by this gene is a member of the tripartite motif (TRIM) family. The TRIM motif includes three zinc-binding domains, a RING, a B-box type 1 and a B-box type 2, and a coiled-coil region. The protein localizes to the cytoplasm. Alternatively spliced transcript variants have been described, but their biological validity has not been determined.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: E3 ubiquitin-protein ligase TRIM15; RING finger protein 93; tripartite motif-containing 15; Zinc finger protein 178; Zinc finger protein B7
Gene Aliases: RNF93; TRIM15; ZNF178; ZNFB7
UniProt ID: (Human) Q9C019
Entrez Gene ID: (Human) 89870
Molecular Function:
ubiquitin-protein ligase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support