Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
Promotions
View available promotion(s)
Promo Code: P5877545 Don't miss out: buy 3, only pay for 2 From June 2-13 take advantage of our 3 for 2 promotion. Whether you are restocking essentials or trying something new, now is a great time to get more for your money!. Learn more
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
FIGURE: 1 / 1
Immunogen sequence: VCRVPGQIEV LRGFLEDVVP DATSAYPYLL LYESRQRRYL GSSPEGSGFC SKDRFVAYPC AVGQTAFSSG RHYWEVGMNI TGDALWALGV CRDNVSRKDR VPKCPENGFW VVQLSKGTKY LSTFSALTPV MLMEPPSHMG IFLDFEAGEV SFYSVSDGSH LHTYSQATFP GPLQPFFCLG APKSGQMVIS TVTMWVKG
The protein encoded by this gene is a member of the tripartite motif (TRIM) family. The TRIM motif includes three zinc-binding domains, a RING, a B-box type 1 and a B-box type 2, and a coiled-coil region. The protein localizes to cytoplasmic bodies. The protein is expressed almost exclusively in the testis, but its function is unknown. Multiple alternatively spliced transcript variants have been found for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: E3 ubiquitin-protein ligase TRIM17; RING finger protein 16; RING finger protein terf; RING-type E3 ubiquitin transferase TRIM17; Testis RING finger protein; tripartite motif protein 17; tripartite motif protein TRIM17; tripartite motif-containing 17; Tripartite motif-containing protein 17
Gene Aliases: RBCC; RNF16; TERF; TRIM17
UniProt ID: (Human) Q9Y577, (Rat) Q9WV59, (Mouse) Q7TPM3
Entrez Gene ID: (Human) 51127, (Rat) 64702, (Mouse) 56631
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support