Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
This Antibody was verified by Cell treatment to ensure that the antibody binds to the antigen stated.
FIGURE: 1 / 9
Recommended positive controls: 293T, A431, H1299, HeLa, HepG2, Molt-4, Raji, TRIM21-transfected 293T.
Predicted reactivity: Cat (86%), Pig (81%), Rhesus Monkey (99%), Bovine (82%).
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
This gene encodes a member of the tripartite motif (TRIM) family. The TRIM motif includes three zinc-binding domains, a RING, a B-box type 1 and a B-box type 2, and a coiled-coil region. The encoded protein is part of the RoSSA ribonucleoprotein, which includes a single polypeptide and one of four small RNA molecules. The RoSSA particle localizes to both the cytoplasm and the nucleus. RoSSA interacts with autoantigens in patients with Sjogren syndrome and systemic lupus erythematosus. Alternatively spliced transcript variants for this gene have been described but the full-length nature of only one has been determined.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 52 kDa ribonucleoprotein autoantigen Ro/SS-A; 52 kDa Ro protein; E3 ubiquitin-protein ligase TRIM21; RING finger protein 81; Ro(SS-A); Sicca syndrome antigen A; Sjoegren syndrome type A antigen; Sjogren syndrome antigen A1 (52kDa, ribonucleoprotein autoantigen SS-A/Ro); SS-A; tripartite motif-containing 21; Tripartite motif-containing protein 21
Gene Aliases: RNF81; Ro/SSA; RO52; SSA; SSA1; TRIM21
UniProt ID: (Human) P19474
Entrez Gene ID: (Human) 6737
Molecular Function:
ubiquitin-protein ligase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support