Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Primary Antibodies
  • Search All
  • Popular Product Areas
  • TaqMan Assays
  • Primary Antibodies
  • Secondary Antibodies
  • Isotype Controls
  • Proteins & Peptides
  • ELISA Kits
  • All Documents & Support
  • Certificates
  • SDS
  • Manuals & Protocols
  • Product FAQs
    Search button Close
            Target of Interest
            Application
            Target Species
            • Order Status
            • Quick Order
            • Sign in
              Sign in
              Don't have an account ? Create Account
              • Account
              • Check Order Status
              • Aspire Member Program
              • Connect: Lab, Data, Apps
              • Custom Products & Projects
              • Services Central

            Disclaimer

            Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

            Loading ...
            • Primary Antibodies ›
            • TRIM23 Antibodies

            Invitrogen

            TRIM23 Polyclonal Antibody

            View all (13) TRIM23 antibodies
            Datasheet
            Protocols
            Questions & Answers
            Datasheet
            Protocols
            Questions & Answers

            Cite TRIM23 Polyclonal Antibody

            • Antibody Testing Data (2)
            Application
            TRIM23 Antibody in Immunocytochemistry (ICC/IF)
            Group 53 Created with Sketch.

            FIGURE: 1 / 2

            TRIM23 Antibody (PA5-97010) in ICC/IF

            Immunofluorescence analysis of TRIM23 in NIH/3T3 cells. Samples were incubated with TRIM23 Polyclonal antibody (Product # PA5-97010) using a dilution of 1:100. Blue: DAPI for nuclear staining.
            View Product

            TRIM23 Antibody in Immunocytochemistry (ICC/IF)
            TRIM23 Antibody in Western Blot (WB)

            Product Details

            PA5-97010

            Applications
            Tested Dilution
            Publications

            Western Blot (WB)

            1:500-1:2,000
            -

            Immunocytochemistry (ICC/IF)

            1:50-1:200
            -

            ELISA (ELISA)

            1 µg/mL
            -
            Product Specifications

            Species Reactivity

            Human, Mouse, Rat

            Host/Isotype

            Rabbit / IgG

            Class

            Polyclonal

            Type

            Antibody

            Immunogen

            Recombinant fusion protein containing a sequence corresponding to amino acids 1-280 of human TRIM23 (NP_0016471)
            View immunogen

            Conjugate

            Unconjugated

            Form

            Liquid

            Concentration

            0.73 mg/mL

            Purification

            Affinity Chromatography

            Storage buffer

            PBS, pH 7.3, with 50% glycerol

            Contains

            0.02% sodium azide

            Storage conditions

            -20°C, Avoid Freeze/Thaw Cycles

            Shipping conditions

            Ambient (domestic); Wet ice (international)

            RRID

            AB_2808812

            Product Specific Information

            Immunogen sequence: MATLVVNKLG AGVDSGRQGS RGTAVVKVLE CGVCEDVFSL QGDKVPRLLL CGHTVCHDCL TRLPLHGRAI RCPFDRQVTD LGDSGVWGLK KNFALLELLE RLQNGPIGQY GAAEESIGIS GESIIRCDED EAHLASVYCT VCATHLCSEC SQVTHSTKTL AKHRRVPLAD KPHEKTMCSQ HQVHAIEFVC LEEGCQTSPL MCCVCKEYGK HQGHKHSVLE PEANQIRASI LDMAHCIRTF TEEISDYSRK LVGIVQHIEG GEQIVEDGIG MAHTEHVPGT; Positive Samples: PC-12, Mouse spleen, Mouse brain; Cellular Location: Endomembrane system, Golgi apparatus membrane, Lysosome membrane

            Target Information

            Acts as an E3 ubiquitin-protein ligase. In the presence of the human cytomegalovirus (HCMV) protein UL144, participates in 'Lys-63'-linked auto-ubiquitination of TRAF6 resulting in the virally controlled activation of NF-kappa-B at early time of infection. The C-terminus can act as an allosteric activator of the cholera toxin catalytic subunit.

            For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

            References (0)

            Have you cited this product in a publication?
            Let us know so we can reference it here.
            Cite this product

            Bioinformatics

            Protein Aliases: ADP-ribosylation factor domain protein 1 64kD; ADP-ribosylation factor domain protein 1, 64kD; ADP-ribosylation factor domain protein 1, 64kDa; ADP-ribosylation factor domain-containing protein 1; ARF domain protein 1; E3 ubiquitin-protein ligase TRIM23; GTP-binding protein ARD-1; nucleotide binding protein; RING finger protein 46; RING-type E3 ubiquitin transferase TRIM23; tripartite motif protein 23; tripartite motif protein TRIM23; tripartite motif-containing 23; Tripartite motif-containing protein 23

            View more View less

            Gene Aliases: 6330516O20Rik; AI450195; Ard-1; ARD1; ARFD1; RNF46; TRIM23

            View more View less

            UniProt ID: (Human) P36406, (Rat) P36407

            View more View less

            Entrez Gene ID: (Human) 373, (Mouse) 81003, (Rat) 81002

            View more View less

            Function(s)

            GTPase activity ubiquitin-protein transferase activity protein binding GTP binding enzyme activator activity zinc ion binding ligase activity GDP binding identical protein binding nucleotide binding UDP-N-acetylmuramoylalanyl-D-glutamyl-2,6-diaminopimelate-D-alanyl-D-alanine ligase activity ribosomal S6-glutamic acid ligase activity coenzyme F420-0 gamma-glutamyl ligase activity coenzyme F420-2 alpha-glutamyl ligase activity metal ion binding protein-glycine ligase activity protein-glycine ligase activity, initiating protein-glycine ligase activity, elongating tubulin-glycine ligase activity protein-glutamic acid ligase activity tubulin-glutamic acid ligase activity G-protein

            Process(es)

            small GTPase mediated signal transduction viral process protein ubiquitination positive regulation of catalytic activity protein ADP-ribosylation
            It has to be done as per old AB suggested Products section.
            guarantee_icon

            Performance Guarantee

            If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

            Learn more
            help_icon

            We're here to help

            Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

            Contact tech support
            Ordering Plus Icon Minus Icon
            • Order Status
            • Order Help
            • Quick Order
            • Supply Center
            • eProcurement
            Support Plus Icon Minus Icon
            • Help and Support
            • Contact Us
            • Technical Support Centers
            • Documents and Certificates
            • Report a Site Issue
            Resources Plus Icon Minus Icon
            • Learning Centers
            • Promotions
            • Events and Webinars
            • Social Media
            About Thermo Fisher Plus Icon Minus Icon
            • About Us
            • Careers
            • Investors
            • News
            • Social Responsibility
            • Trademarks
            • Consumer Health Data Privacy Policy
            Our Portfolio Plus Icon Minus Icon
            • Thermo Scientific
            • Applied Biosystems
            • Invitrogen
            • Gibco
            • Ion Torrent
            • Fisher Scientific
            • Unity Lab Services
            • Patheon
            • PPD
            • Terms & Conditions
            • Privacy Information Center
            • Price & Freight Policy
            • Cookie Preferences

              Your choices regarding cookies on this site

              We and our affiliates and vendors use cookies and similar technologies to operate our sites, recognize visitors to our sites, provide secure log-in, collect statistics to optimize site functionality, and deliver content tailored to your interests. Click Accept All to accept all cookies and go directly to the site, click Reject All to reject all but cookies strictly necessary to the functioning of this site (required cookies), or click on Manage Settings to see detailed descriptions of the types of cookies and choose whether to accept certain cookies while on the site.
            © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
            United States flag icon
            United States

            Your items have has been added!


            Host server : magellan-search-green-56756dfb8b-mxnmt:80/100.66.74.71:80.
            git-commit: de6905f249d25e22729e613aef254bb544bd8b7c
            git-url: https://github.com/thermofisher/magellan-search
            git-branch: release/2.31.1-2025.06.25-1.0