Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: NHLSTLRRD RDKIQGFQAK GEADILAALK KLQDQRQYIV AEFEQGHQFL REREEHLLEQ LAKLEQELTE GREKFKSRGV GELARLALVI SELEGKAQQP AAELMQDTRD FLNRYPRKKF W (151-270 aa encoded by BC024039)
TRIM26 is a member of the tripartite motif (TRIM) family. The TRIM motif includes three zinc-binding domains, a RING, a B-box type 1 and a B-box type 2, and a coiled-coil region. The protein localizes to cytoplasmic bodies. Although the function of the protein is unknown, the RING domain suggests that the protein may have DNA-binding activity. The protein encoded by this gene is a member of the tripartite motif (TRIM) family. The TRIM motif includes three zinc-binding domains, a RING, a B-box type 1 and a B-box type 2, and a coiled-coil region. The protein localizes to cytoplasmic bodies. Although the function of the protein is unknown, the RING domain suggests that the protein may have DNA-binding activity. The gene localizes to the major histocompatibility complex (MHC) class I region on chromosome 6.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Acid finger protein; AFP; RING finger protein 95; tripartite motif protein 26; tripartite motif protein TRIM26; tripartite motif-containing 26; Tripartite motif-containing protein 26; widely expressed acid zinc finger protein; Zinc finger protein 173
Gene Aliases: AFP; AI462198; RNF95; TRIM26; Zfp173; Zfp1736; ZNF173
UniProt ID: (Human) Q12899, (Mouse) Q99PN3
Entrez Gene ID: (Human) 7726, (Mouse) 22670
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support