Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
Promotions
View available promotion(s)
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
FIGURE: 1 / 7
Recommended positive controls: 293T, DDDDK-TRIM32-transfected 293T.
Predicted reactivity: Mouse (97%), Rat (98%), Cat (98%), Rhesus Monkey (99%), Bovine (91%).
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
TRIM32 is a member of the tripartite motif (TRIM) family. The TRIM motif includes three zinc-binding domains, a RING, a B-box type 1 and a B-box type 2, and a coiled-coil region. TRIM32 localizes to cytoplasmic bodies. The protein has also been localized to the nucleus, where it interacts with the activation domain of the HIV-1 Tat protein. The Tat protein activates transcription of HIV-1 genes.
⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 72 kDa Tat-interacting protein; bM468K2.2 (tripartite motif protein 32); E3 ubiquitin-protein ligase TRIM32; RING-type E3 ubiquitin transferase TRIM32; RP11-67K19.1; TAT-interactive protein, 72-KD; tripartite motif protein 32; tripartite motif-containing 32; Tripartite motif-containing protein 32; zinc finger protein 117; Zinc finger protein HT2A; zinc-finger protein HT2A
Gene Aliases: 1810045E12Rik; 3f3; BBS11; HT2A; LGMD2H; TATIP; TRIM32; Zfp117
UniProt ID: (Human) Q13049, (Mouse) Q8CH72
Entrez Gene ID: (Human) 22954, (Mouse) 69807
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support