Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MAEGEDMQTF TSIMDALVRI STSMKNMEKE LLCPVCQEMY KQPLVLPCTH NVCQACAREV LGQQGYIGHG GDPSSEPTSP ASTPSTRSPR LSRRTLPKPD RLDRLLKSGF GTYPGRKRGA LHPQVIMFPC PACQGDVELG ERGLAGLFRN LTLERVVERY RQS
Microtubule-associated protein that is involved in the formation of parallel microtubule bundles linked by cross-bridges in the proximal axon. Required for the uniform orientation and maintenance of the parallel microtubule fascicles, which are important for efficient cargo delivery and trafficking in axons. Thereby also required for proper axon specification, the establishment of neuronal polarity and proper neuronal migration. [UniProt]
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Gene Y protein; GeneY; tripartite motif protein 46; Tripartite motif-containing protein 46; tripartite, fibronectin type III and C-terminal SPRY motif-containing; Tripartite, fibronectin type-III and C-terminal SPRY motif protein
Gene Aliases: GENEY; TRIFIC; TRIM46
UniProt ID: (Human) Q7Z4K8, (Mouse) Q7TNM2
Entrez Gene ID: (Human) 80128, (Mouse) 360213, (Rat) 310641
Molecular Function: ubiquitin-protein ligase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support