Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MCPDHKEEVT HYCKTCQRLV CQLCRVRRTH SGHKITPVLS AYQALKDKLT KSLTYILGNQ DTVQTQICEL EEAVRHTEVS GQQAKEEVSQ LVRGLGAVLE EKR
Highest antigen sequence identity to the following orthologs: Mouse - 97%, Rat - 97%.
Microtubule-associated protein that is involved in the formation of parallel microtubule bundles linked by cross-bridges in the proximal axon. Required for the uniform orientation and maintenance of the parallel microtubule fascicles, which are important for efficient cargo delivery and trafficking in axons. Thereby also required for proper axon specification, the establishment of neuronal polarity and proper neuronal migration. [UniProt]
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Gene Y protein; GeneY; Tripartite motif-containing protein 46; Tripartite, fibronectin type-III and C-terminal SPRY motif protein
Gene Aliases: GENEY; TRIFIC; TRIM46
UniProt ID: (Human) Q7Z4K8
Entrez Gene ID: (Human) 80128
Molecular Function: ubiquitin-protein ligase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support