Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: QGLWLLGLRE GKILEAHVEA KEPRALRSPE RRPTRIGLYL SFGDGVLSFY DASDADALVP LFAFHERLPR PVYPFFDVCW HDKGKNAQPL LLVGPEGAEA
Muscle-specific protein that plays a central role in cell membrane repair by nucleating the assembly of the repair machinery at injury sites. Specifically binds phosphatidylserine. Acts as a sensor of oxidation: upon membrane damage, entry of extracellular oxidative environment results in disulfide bond formation and homooligomerization at the injury site. This oligomerization acts as a nucleation site for recruitment of TRIM72-containing vesicles to the injury site, leading to membrane patch formation. Probably acts upstream of the Ca(2+)-dependent membrane resealing process. Required for transport of DYSF to sites of cell injury during repair patch formation. Regulates membrane budding and exocytosis. May be involved in the regulation of the mobility of KCNB1-containing endocytic vesicles.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Mg53; mitsugumin 53; Mitsugumin-53; TRIM72; tripartite motif containing 72, E3 ubiquitin protein ligase; tripartite motif-containing 72; Tripartite motif-containing protein 72
Gene Aliases: MG53; TRIM72
UniProt ID: (Human) Q6ZMU5
Entrez Gene ID: (Human) 493829
Molecular Function:
ubiquitin-protein ligase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support