Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
Promotions
View available promotion(s)
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
Recommended positive controls: HepG2, 3XFlag-human TRIM8-transfected 293T.
Predicted reactivity: Mouse (98%), Rat (98%), Xenopus laevis (91%), Cat (100%), Chicken (94%), Rhesus Monkey (100%), Bovine (100%).
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
The protein encoded by this gene is a member of the tripartite motif (TRIM) family. The TRIM motif includes three zinc-binding domains, a RING, a B-box type 1 and a B-box type 2, and a coiled-coil region. This protein localizes to nuclear bodies. Its structure is similar to some tumor suppressor proteins and its gene maps to a locus thought to contain tumor suppressor genes.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: E3 ubiquitin-protein ligase TRIM8; glioblastoma expressed ring finger protein; Glioblastoma-expressed RING finger protein; RING finger protein 27; RING-type E3 ubiquitin transferase TRIM8; tripartite motif protein TRIM8; tripartite motif-containing 8; Tripartite motif-containing protein 8
Gene Aliases: GERP; RNF27; TRIM8
UniProt ID: (Human) Q9BZR9
Entrez Gene ID: (Human) 81603
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support