Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: VIDDESDYF ASDSNQWLSK LERETLQKRE EELRELRHAS RLSKKVTIDF AGRKILEEEN SLAEYHSRLD ETIQAIANGT LNQPLTKLDR SSEEPLGVLV NPNMYQSPPQ WVDHTGAASQ KKAFRSSGFG LEFNSFQHQL RIQDQEFQEG FDGGWCLSVH QPWASLLVRG IKRVEGRSWY TPHRGRLWIA ATAKKPSPQE VSELQATYRL LRGKDVEFPN DYPSGCLLGC VDLIDCLSQK QFKEQFPDIS QESDSPFVFI CKNPQEMVVK FPIKGNPKIW KLDSKIHQGA KKGLMKQNKA V (282-581 aa encoded by BC012448)
Transcription coactivator of nuclear receptors which functions in conjunction with CBP-p300 and SRC-1 and may play an important role in establishing distinct coactivator complexes under different cellular conditions. Plays a pivotal role in the transactivation of NF-kappa-B, SRF and AP1. Acts as a mediator of transrepression between nuclear receptor and either AP1 or NF-kappa-B. Plays a role in androgen receptor transactivation and in testicular function.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Activating signal cointegrator 1; ASC-1; Thyroid receptor-interacting protein 4; TR-interacting protein 4; transcription coactivator Asc1; TRIP-4; zinc finger, C2HC5-type
Gene Aliases: 4930558E03Rik; ASC-1; ASC1; BB191711; HsT17391; RQT4; SMABF1; TRIP4; ZC2HC5
UniProt ID: (Human) Q15650, (Mouse) Q9QXN3
Entrez Gene ID: (Human) 9325, (Rat) 315769, (Mouse) 56404
Molecular Function:
transcription cofactor
gene-specific transcriptional regulator
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support