Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
Promotions
View available promotion(s)
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
Immunogen sequence: PPPAYRTGS LKPNPASPLP ASPYGGPTPA SYTTASTPAG PAFPVQVKVA QPVRGCGPPR RGASQASGPL PGPHFPLPGR GEVWGPGYRS QREPGPGAKE EAAGVSGPAG RGRGGEHGPQ VPLSQPPEDE LDRLTKKLVH DMNHPPSGEY FGQCGGCGED VVGDGAGVVA FDRVFHVGCF VCSTCRAQLR GQHFYAVERR AYCEGCYVAT LEKCATCSQP ILDRILRAMG KAYHPGCFTC VVCHRGLDGI PFTVDATSQI HCIEDFHRKF APRCSVCGGA IMPEPGQEET VRIVALDRSF HIGCYKCEEC GLLLSSEGEC QGCYPLDGHI LCKACSAWRI QELSATVTTD C (127-476 aa encoded by BC004249)
This gene is a member of the zyxin family and encodes a protein with three LIM zinc-binding domains. This protein localizes to focal adhesion sites and along actin stress fibers. Recruitment of this protein to the plasma membrane occurs in a lysophosphatidic acid (LPA)-dependent manner and it regulates LPA-induced cell migration. Alternatively spliced variants which encode different protein isoforms have been described; however, not all variants have been fully characterized.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: MGC10556; MGC10558; MGC29959; MGC3837; MGC4423; OIP-1; OPA-interacting protein 1; tcag7.1300; thyroid hormone receptor interacting protein 6; Thyroid receptor-interacting protein 6; TR-interacting protein 6; TRIP-6; ZRP-1; zyxin related protein 1; Zyxin-related protein 1
Gene Aliases: OIP-1; OIP1; TRIP-6; TRIP6; TRIP6i2; ZRP-1; Zrp1
UniProt ID: (Human) Q15654, (Mouse) Q9Z1Y4
Entrez Gene ID: (Human) 7205, (Mouse) 22051
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support