Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
Promotions
View available promotion(s)
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
Immunogen sequence: EHCGQRHQLG GPMWAEPIHD LDFVGRVLEA VSANPGRFHT SERIRGVLSV ITEELPDVPL YYTLDQLSST IHCNTP
Highest antigen sequence identity to the following orthologs: Mouse - 92%, Rat - 91%.
TRMT1 is a tRNA(m(2)(2)G(26))dimethyltransferase.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: FLJ20244; N(2),N(2)-dimethylguanosine tRNA methyltransferase; TRM1 tRNA methyltransferase 1 homolog; tRNA (guanine(26)-N(2))-dimethyltransferase; tRNA 2,2-dimethylguanosine-26 methyltransferase; tRNA methyltransferase 1 homolog; tRNA(guanine-26,N(2)-N(2)) methyltransferase; tRNA(m(2,2)G26)dimethyltransferase
Gene Aliases: TRM1; TRMT1
UniProt ID: (Human) Q9NXH9
Entrez Gene ID: (Human) 55621
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support