Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: TPLWRLSYE EQLKVKFAAQ KKILQRLESY IQMLNGVSVT TAVPKSERLS CLLHPIIPSP VINGYRNKST FSVNRGPDGN PKTVGFYLGT WRDGNVVCVQ SNHLKNIPEK HSQVAQYYEV FLRQSPLEPC LVFHEGGYWR ELTVRTNSQG HTMAIITFHP (89-247 aa encoded by BC007526)
Probable S-adenosyl-L-methionine-dependent methyltransferase that catalyzes the formation of 5-methyl-uridine at position 54 (m5U54) in all tRNA. May also have a role in tRNA stabilization or maturation.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: rRNA (uracil-5-)-methyltransferase TRMT2B; TRM2 homolog; TRM2 homolog B; TRM2 tRNA methyltransferase 2 homolog B; tRNA (uracil-5-)-methyltransferase homolog; tRNA (uracil-5-)-methyltransferase homolog B
Gene Aliases: CXorf34; dJ341D10.3; TRMT2B
UniProt ID: (Human) Q96GJ1
Entrez Gene ID: (Human) 79979
Molecular Function: RNA methyltransferase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support