Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
The protein encoded by this gene is similar to the transient receptor potential (Trp) calcium channel family members. The expression of this protein is inversely correlated with melanoma aggressiveness, suggesting that it suppresses melanoma metastasis.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Long transient receptor potential channel 1; LTrpC1; melastatin 1; Melastatin-1; Transient receptor potential cation channel subfamily M member 1; transient receptor potential cation channel, subfamily M, member 1; transient receptor potential melastatin family
Gene Aliases: CSNB1C; LTRPC1; MLSN; MLSN1; TRPM1
UniProt ID: (Human) Q7Z4N2
Entrez Gene ID: (Human) 4308
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support