Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TRPM8 is a receptor-activated non-selective cation channel involved in detection of sensations such as coolness, by being activated by cold temperature below 25 degrees Celsius. TRPM8 is activated by icilin, eucalyptol, menthol, cold and modulation of intracellular pH and involved in menthol sensation. TRPM8 is permeable for monovalent cations sodium, potassium, and cesium and divalent cation calcium. TRPM8 has an expression pattern within normal human tissues that is mostly restricted to prostate epithelial cells. TRPM8 mRNA expression is at moderate levels in normal prostate tissue and appears to be elevated in prostate cancer. Notably, TRPM8 mRNA was also expressed in a number of nonprostatic primary tumors of breast, colon, lung, and skin origin, whereas transcripts encoding TRPM8 were hardly detected or not detected in the corresponding normal human tissues.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Cold menthol receptor 1; cold/menthol receptor 1; Long transient receptor potential channel 6; LTrpC-6; menthol receptor; MGC2849; short form of the TRPM8 cationic channel; transient potential cation channel subfamily M member 8; transient receptor melastatin 8 variant 1; transient receptor melastatin 8 variant 2; Transient receptor potential cation channel subfamily M member 8; transient receptor potential cation channel, subfamily M, member 8; transient receptor potential cation channel, subfamily M, member 8 (predicted), 3 prime; transient receptor potential cation channel, subfamily M, member 8 (predicted), 5 prime; transient receptor potential M8 protein; Transient receptor potential p8; transient receptor potential subfamily M member 8; transient receptor potential-p8; TRP ion channel TRPM8; Trp-p8
Gene Aliases: CMR1; I79_024425; LTrpC-6; LTRPC6; Trp-p8; TRPM8; TRPP8
UniProt ID: (Human) Q7Z2W7, (Rat) Q8R455, (Mouse) Q8R4D5
Entrez Gene ID: (Rabbit) 100351107, (Human) 79054, (Horse) 100057419, (Chicken) 424025, (Dog) 486170, (Rat) 171384, (Guinea pig) 100337634, (Chinese hamster) 100754189, (Mouse) 171382
Molecular Function:
ion channel
transporter
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support