Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
Promotions
View available promotion(s)
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
Peptide sequence: RVDEVNWSHW NQNLGIINED PGKNETYQYY GFSHTVGRLR RDRWSSVVPR
Sequence homology: Cow: 93%; Dog: 86%; Guinea Pig: 86%; Human: 100%; Mouse: 86%; Pig: 93%; Rabbit: 100%; Rat: 93%
TRPV4 (Transient Receptor Potential Cation Channel Subfamily V Member 4) is a cation-selective channel, activated in response to systemic osmotic pressure. Non-selective calcium permeant cation channel probably involved in osmotic sensitivity and mechanosensitivity. TRPV4 is a member of the OSM9-like transient receptor potential channel (OTRPC) subfamily in the transient receptor potential (TRP) superfamily of ion channels. The TRPV4 protein is a Ca2+-permeable, nonselective cation channel that is thought to be involved in the regulation of systemic osmotic pressure. Mutations in the TRPV4 gene are the cause of spondylometaphyseal and metatropic dysplasia and hereditary motor and sensory neuropathy type IIC. Diseases associated with TRPV4 include Metatropic Dysplasia and Spondylometaphyseal Dysplasia, Kozlowski Type. Multiple transcript variants encoding different isoforms have been found for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Osm-9-like TRP channel 4; OSM9-like transient receptor potential channel 4; osmosensitive transient receptor potential channel 4; OTRPC4; Transient receptor potential cation channel subfamily V member 4; transient receptor potential cation channel, subfamily 5, member 4; transient receptor potential cation channel, subfamily V, member 4; Transient receptor potential protein 12; TRP12; TrpV4; Vanilloi; Vanilloid receptor-like channel 2; Vanilloid receptor-like protein 2; vanilloid receptor-related osmotically activated channel; vanilloid receptor-related osmotically activated channel (Vroac); Vanilloid receptor-related osmotically-activated channel; VR-OAC; VRL-2
Gene Aliases: BCYM3; CMT2C; HMSN2C; OTRPC4; SMAL; SPSMA; SSQTL1; TRP12; TRPV4; VRL2; VROAC
UniProt ID: (Human) Q9HBA0, (Rat) Q9ERZ8
Entrez Gene ID: (Human) 59341, (Rat) 66026
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support