Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MAASEAAVV SSPSLKTDTS PVLETAGTVA AMAATPSARA AAAVVAAAAR TGSEARVSKA ALATKLLSLS GVFAVHKPKG PTSAELLNRL KEKLLAEAGM PSPEWTKRKK QTLKIGHGGT LDSAARGVLV VGIGSGTKML TSMLSGSKRY TAIGELGKAT DTLDSTGRVT EEKPYDKITQ EDIEGILQKF TGNIMQVPPL YSALKKDGQR LSTLMKRGEV VEAKPARPVT VYSISLQKFQ PPFFTLDVEC GGGFYIRSLV SDIGKELSSC ANVLELTRTK QGPFTLEEHA LPEDKWTIDD IAQSLEHCSS LFPAELALKK SKPESNEQVL SCEYITLNEP KREDDVIKTC (1-349 aa encoded by BC030601)
Pseudouridine is an abundant component of rRNAs and tRNAs and is enzymatically generated by isomerization of uridine by pseudouridine synthase (Zucchini et al., 2003 ).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: probable tRNA pseudouridine synthase 1; Pseudouridylate synthase TRUB1; Psi55 synthase TRUB1; tRNA pseudouridine 55 synthase TRUB1; TruB pseudouridine (psi) synthase family member 1; TruB pseudouridine (psi) synthase homolog 1; TruB pseudouridine synthase homolog 1
Gene Aliases: 2610009I02Rik; 9030425C13Rik; AI448235; BB118943; C76870; PUS4; TRUB1
UniProt ID: (Human) Q8WWH5, (Mouse) Q8C0D0
Entrez Gene ID: (Human) 142940, (Mouse) 72133
Molecular Function:
isomerase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support