Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MKFILLWALL NLTVALAFNP DYTVSSTPPY LVYLKSDYLP CAGVLIHPLW VITAAHCNLP KLRVILGVTI PADSNEKHLQ VIGYEKMIHH PHFSVTSIDH DIMLIKLKTE AELNDYVKLA NLPYQTISEN TMCSVSTWSY NVCDIYKEPD SLQTVNISVI SKPQCRDAYK TYNITENMLC VGIVPGRRQP CKEVSAAPAI CNGMLQGILS FADGCVLRAD VGIYAKIFYY IPWIENVIQN N
This gene encodes a member of the trypsin family of serine proteases. This gene and several related trypsinogen genes are localized to the T cell receptor beta locus on chromosome 7. This gene was previously described as a trypsinogen-like pseudogene, but it is now thought to be a protein-coding gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: protease, serine, 58; Serine protease 58; Trypsin-X3
Gene Aliases: PRSS1; PRSS58; TRY1; TRYX3; UNQ2540; UNQ2540/PRO6090
UniProt ID: (Human) Q8IYP2
Entrez Gene ID: (Human) 136541
Molecular Function:
protease
serine protease
protein modifying enzyme
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support