Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: KTNMPIITSK RYQHSVEWAA ELMRRQGQDE STVRRILKDY TKPLEHPPVK RNEEAQVHDK LNSGMVSNME GTAGGERPSV VNGDSGKSGG VGDPREP
Highest antigen sequence identity to the following orthologs: Mouse - 45%, Rat - 48%.
Constitutes one of the two catalytic subunit of the tRNA-splicing endonuclease complex, a complex responsible for identification and cleavage of the splice sites in pre-tRNA. It cleaves pre-tRNA at the 5'- and 3'-splice sites to release the intron. The products are an intron and two tRNA half-molecules bearing 2',3'-cyclic phosphate and 5'-OH termini. There are no conserved sequences at the splice sites, but the intron is invariably located at the same site in the gene, placing the splice sites an invariant distance from the constant structural features of the tRNA body. Isoform 1 probably carries the active site for 5'-splice site cleavage. The tRNA splicing endonuclease is also involved in mRNA processing via its association with pre- mRNA 3'-end processing factors, establishing a link between pre- tRNA splicing and pre-mRNA 3'-end formation, suggesting that the endonuclease subunits function in multiple RNA-processing events. Isoform 2 is responsible for processing a yet unknown RNA substrate. The complex containing isoform 2 is not able to cleave pre-tRNAs properly, although it retains endonucleolytic activity.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: HsSen2; tRNA-intron endonuclease Sen2; tRNA-splicing endonuclease subunit Sen2; TSEN2 tRNA splicing endonuclease subunit
Gene Aliases: PCH2B; SEN2; SEN2L; TSEN2
UniProt ID: (Human) Q8NCE0
Entrez Gene ID: (Human) 80746
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support