Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 2
Sequence of this protein is as follows: MSLLRSLRVF LVARTGSYPA GSLLRQSPQP RHTFYAGPRL SASASSKELL MKLRRKTGYS FVNCKKALET CGGDLKQAEI WLHKEAQKEG WSKAAKLQGR KTKEGLIGLL QEGNTTVLVE VNCETDFVSR NLKFQLLVQQ VALGTMMHCQ TLKDQPSAYS KVQWLTPVNL ALWEAEAGGS LEGFLNSSEL SGLPAGPDRE GSLKDQLALA IGKLGENMIL KRAAWVKVPS GFYVGSYVHG AMQSPSLHKL VLGKYGALVI CETSEQKTNL EDVGRRLGQH VVGMAPLSVG SLDDEPGGEA ETKMLSQPYL LDPSITLGQY VQPQGVSVVD FVRFECGEGE EAAETE
Synthesis of the 13 mitochondrial-encoded proteins occurs on a dedicated mitochondrial translation apparatus similar to that found in prokaryotes and requires, in addition to the tRNAs and rRNAs encoded in mtDNA, the concerted action of several translation factors and a large number of mitochondrial ribosomal proteins, all of which are encoded by nuclear genes. The TSFM gene encodes a mitochondrial translation elongation factor (Smeitink et al., 2006 ).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: EF-Ts; EF-TsMt; Elongation factor Ts, mitochondrial; mitochondrial elongation factor Ts; TSFM
Gene Aliases: EFTS; EFTSMT; TSFM
UniProt ID: (Human) P43897
Entrez Gene ID: (Human) 10102
Molecular Function:
translation elongation factor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support