Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Sandwich ELISA (Recombinant protein).
Immunogen sequence: KVQWLTPVNL ALWEAEAGGS LEGFLNSSEL SGLPAGPDRE GSLKDQLALA IGKLGENMIL KRAAWVKVPS GFYVGSYVHG AMQSPSLHKL VLGKYGALVI
Synthesis of the 13 mitochondrial-encoded proteins occurs on a dedicated mitochondrial translation apparatus similar to that found in prokaryotes and requires, in addition to the tRNAs and rRNAs encoded in mtDNA, the concerted action of several translation factors and a large number of mitochondrial ribosomal proteins, all of which are encoded by nuclear genes. The TSFM gene encodes a mitochondrial translation elongation factor (Smeitink et al., 2006 ).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: EF-Ts; EF-TsMt; Elongation factor Ts, mitochondrial; mitochondrial elongation factor Ts; TSFM
Gene Aliases: EFTS; EFTSMT; TSFM
UniProt ID: (Human) P43897
Entrez Gene ID: (Human) 10102
Molecular Function:
translation elongation factor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support