Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
Immunogen sequence: GPSRDGTISE DTIRASLISA VSDKLRWRMK EEMDRAQAEL NALKRTEEDL KKGHQKLEEM VTRLDQEVAE VDKNIELLKK KDEELSSALE KMENQSENND IDEVIIPTAP LYKQILNLYA EENAIEDTIL YLGEALRRGV IDLDVFLKHV RLLSRKQFQL RALMQKARKT AGLSDLY
TSG101-4A10 recognizes TSG101 protein, the product of a recently identified Tumor Susceptibility Gene whose inactivation in mouse fibroblasts results in cell transformation and the ability of those cells to form tumors in nude mice. The TSG101 gene encodes a 46 kDa protein and contains a proline-rich domain and a leucine-zipper motif within a coiled-coil domain near the carboxyl terminus.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ESCRT-I complex subunit TSG101; ESCRTI complex subunit TSG101; TSG 101; Tumor supressor; tumor susceptibility gene 10; tumor susceptibility gene 101; Tumor susceptibility gene 101 protein; tumor susceptibility protein
Gene Aliases: AI255943; CC2; Rw; TSG10; TSG101; VPS23
UniProt ID: (Human) Q99816, (Rat) Q6IRE4, (Mouse) Q61187
Entrez Gene ID: (Human) 7251, (Rat) 292925, (Mouse) 22088
Molecular Function: ubiquitin-protein ligase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support