Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
MA1-82909 is specific for beta (TSH) and shows minimal cross-reactivity with beta Human chorionic gonadotropin (hCG), Luteinizing Hormone (LH) or Follicle Stimulating Hormone (FSH).
Mouse anti Human thyroid stimulating hormone, clone 155 recognizes the beta chain of human beta (TSH) and shows minimal cross-reactivity with human chorionic gonadotropin (hCG), luteinizing hormone (hLH) or follicle stimulating hormone (hFSH).
TSH is a pituitary hormone of 28kDa, which stimulates thyroid growth and production of thyroid hormones. This antibody labels thyrotropic cells of the pituitary.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: thyroid stimulating hormone, beta; Thyroid-stimulating hormone subunit beta; Thyrotropin beta chain; Thyrotropin subunit beta; TSH; TSH-B
Gene Aliases: TSH-B; TSH-BETA; TSHB
UniProt ID: (Human) P01222
Entrez Gene ID: (Human) 7252
Molecular Function: peptide hormone
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support