Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
Promotions
View available promotion(s)
Promo Code: P5877545 Don't miss out: buy 3, only pay for 2 From June 2-13 take advantage of our 3 for 2 promotion. Whether you are restocking essentials or trying something new, now is a great time to get more for your money!. Learn more
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
This rabbit polyclonal antibody raised against N-terminal region (1-253 aa) of human TSHR can detect subunit A with an experimentally determined molecular weight of 62-70 kDa under reducing condition.
Immunogen sequence: MRPADLLQL VLLLDLPRDL GGMGCSSPPC ECHQEEDFRV TCKDIQRIPS LPPSTQTLKL IETHLRTIPS HAFSNLPNIS RIYVSIDVTL QQLESHSFYN LSKVTHIEIR NTRNLTYIDP DALKELPLLK FLGIFNTGLK MFPDLTKVYS TDIFFILEIT DNPYMTSIPV NAFQGLCNET LTLKLYNNGF TSVQGYAFNG TKLDAVYLNK NKYLTVIDKD AFGGVYSGPS LLLPLGRKSL SFETQKAPRS SMPS (1-253 aa encoded by BC063613)
TSH Receptor gene is a membrane protein and a major controller of thyroid cell metabolism. The encoded protein is a receptor for thyrothropin and thyrostimulin, and its activity is mediated by adenylate cyclase. Defects in this gene are a cause of several types of hyperthyroidism. Three transcript variants encoding different isoforms have been found for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: MGC141920; seven transmembrane helix receptor; thyroid stimulating hormone receptor, isoform 2; Thyroid-stimulating hormone receptor; Thyrotropin receptor; thyrotropin receptor-I, hTSHR-I; TSH-R
Gene Aliases: AI481368; CHNG1; hTSHR-I; hypothroid; hyt; LGR3; pet; TSHR; TSHRA
UniProt ID: (Human) P16473, (Mouse) P47750, (Rat) P21463
Entrez Gene ID: (Human) 7253, (Mouse) 22095, (Rat) 25360
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support