Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
A suggested positive control is mouse heart tissue lysate.
PA5-20380 can be used with blocking peptide PEP-0497.
Thymic stromal lymphopoietin (TSLP) has recently been identified as an important factor capable of driving dendritic cell maturation and activation. It is involved in the positive selection of regulatory T cells, maintenance of peripheral CD4+ T cell homeostasis and the induction of CD4+ T cell-mediated allergic reaction. TSLP is also capable of supporting the growth of fetal liver and adult B cell progenitors and their differentiation to the IgM-positive stage of B cell development. Its receptor TSLP-R will bind TSLP in a low-affinity fashion in transfected cells; co-transfection with IL-7R-alpha cDNA results in high-affinity binding and a functional heteromeric complex. This heteromeric receptor requires stat5 for TSLP-mediated signal transduction and is inhibited by SOCS-1.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CRLM-2; Cytokine receptor-like factor 2; Cytokine receptor-like molecule 2; delta 1; lymphocyte antigen 114; P2RY8/CRLF2 fusion; Thymic stromal lymphopoietin protein receptor; thymic stromal-derived lymphopoietin, receptor; transmembrane phosphoinositide 3-phosphatase and tensin homolog 2; TSLP receptor; Type I cytokine receptor delta 1; WI2-87105E3.1
Gene Aliases: Crlf2; CRLM2; Ly114; Tpte2; Tslpr
UniProt ID: (Mouse) Q8CII9
Entrez Gene ID: (Mouse) 57914
Molecular Function: transmembrane signal receptor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support