Recommended positive controls: A549, H1299, HCT116, MCF-7.
Predicted reactivity: Mouse (84%), Rat (85%), Rhesus Monkey (97%), Bovine (87%).
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
May be involved in 20S pre-rRNA processing.
⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Pre-rRNA-processing protein TSR2 homolog; TSR2, 20S rRNA accumulation, homolog; WGG motif containing 1
Gene Aliases: DBA14; DT1P1A10; TSR2; WGG1
UniProt ID: (Human) Q969E8
Entrez Gene ID: (Human) 90121
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support