Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
Promotions
View available promotion(s)
Promo Code: P5877545 Don't miss out: buy 3, only pay for 2 Extended to June 20: take advantage of our 3 for 2 promotion. Whether you are restocking essentials or trying something new, now is a great time to get more for your money!. Learn more
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
Immunogen sequence: DKWENVKISD FGFAKMVPSN QPVGCSPSYR QVNCFSHLSQ TYCGSFAYAC PEILRGLPYN PFLSDTWSMG VILYTLVVAH LPFDDTNLKK LLRETQKEVT FPANHTISQE CKNLILQMLR QATKRATILD IIKDSWVLKF QPEQPTHEIR LLEAMCQLHN TTKQHQSLQI TT
TSSK4 belongs to a family of serine/threonine kinases highly expressed in testis (Chen et al., 2005 [PubMed 15964553]).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: serine/threonine kinase 22E; Serine/threonine-protein kinase 22E; testis-specific kinase 4; testis-specific serine kinase 4; testis-specific serine/threonine protein kinase 5 variant alpha; testis-specific serine/threonine protein kinase 5 variant beta; testis-specific serine/threonine protein kinase 5 variant delta; testis-specific serine/threonine protein kinase 5 variant gamma; Testis-specific serine/threonine-protein kinase 4; TSK-4
Gene Aliases: 1700020B19Rik; 4933424F08Rik; C14orf20; RGD1560814; STK22E; TSK-4; TSK4; TSSK-4; TSSK4; TSSK5
UniProt ID: (Human) Q6SA08, (Mouse) A3QQR1
Entrez Gene ID: (Human) 283629, (Mouse) 71099, (Rat) 290229
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support