Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Tau tubulin kinase (TTBK2) is a serine/threonine kinase whose substrates are thought to include the tau and tubulin proteins. Mutations in the TTBK2 gene cause spinocerebellar ataxia type 11 (SCA11), a neurodegenerative disease characterized by progressive ataxia and atrophy of the cerebellum and brainstem. Recent studies have shown that increased levels of TTBK2 expression in kidney carcinoma and melanoma cell lines increases resistance to the chemotherapeutic agent Sunitinib, and that reduction of TTBK2 though the use of siRNA sensitized these cell lines to the effects of Sunitib, suggesting that TTBK2 can be used as a resistance marker as well a potential target to overcome resistance.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Protein bartleby; Tau-tubulin kinase 2
Gene Aliases: 2610507N02Rik; AI326283; B930008N24Rik; Bby; KIAA0847; mKIAA0847; SCA11; TTBK; Ttbk1; TTBK2; TTK
UniProt ID: (Human) Q6IQ55, (Mouse) Q3UVR3
Entrez Gene ID: (Human) 146057, (Mouse) 140810
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support