Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Recommended positive controls: NT2D1, IMR32, U87-MG.
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
TTC14 is a protein coding gene.
⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Tetratricopeptide repeat protein 14; TPR repeat protein 14; UNQ5813
Gene Aliases: DRDL5813; KIAA1980; PRO19630; TTC14; UNQ5813/PRO19630
UniProt ID: (Human) Q96N46
Entrez Gene ID: (Human) 151613
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support