Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
The recommended shelf life for this product is 1 year from date of receipt.
TTC33 is a protein coding gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: osmosis responsive factor; Osmosis-responsive factor; Tetratricopeptide repeat protein 33; TPR repeat protein 33
Gene Aliases: 2410099M07Rik; 2900001O04Rik; AI507072; AW538342; OSRF; TTC33
UniProt ID: (Human) Q6PID6, (Mouse) Q9D6K7
Entrez Gene ID: (Human) 23548, (Mouse) 67515
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support