Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MESEDLSGRE LTIDSIMNKV RDIKNKFKNE DLTDELSLNK ISADTTDNSG TVNQIMMMAN NPEDWLSLLL KLEKNSVPLS DALLNKLIGR YSQAIEALPP DKYGQNESFA RIQVRFAELK AIQEPDDARD YFQMARANCK KFAFVHISFA QFELSQGNVK KSKQLLQKAV ERGAVPLEML EIALRNLNLQ KKQLLSEEEK KNLSASTVLT AQESFSGSLG HLQNRNNSCD SRGQTTKARF LYGENMPPQD AEIGYRNSLR QTNKTKQSCP FGRVPVNLLN SPDCDVKTDD SVVPCFMKRQ TSRSECRDLV VPGSKPSGND SCELRNLKSV QNSHFKEPLV SDEKSSELII TDSITLKNKT ESSLLAKLEE TKEYQEPEVP ESNQKQWQSK RKSECINQNP AASSNHWQIP ELARKVNTEQ KHTTFEQPVF SVSKQSPPIS TSKWFDPKSI CKTPSSNTLD DYMSCFRTPV VKNDFPPACQ LSTPYGQPAC FQQQQHQILA TPLQNLQVLA SSSANECISV KGRIYSILKQ IGSGGSSKVF QVLNEKKQIY AIKYVNLEEA DNQTLDSYRN EIAYLNKLQQ HSDKIIRLYD YEITDQYIYM VMECGNIDLN SWLKKKKSID PWERKSYWKN MLEAVHTIHQ HGIVHSDLKP ANFLIVDGML KLIDFGIANQ MQPDTTSVVK DSQVGTVNYM PPEAIKDMSS SRENGKSKSK ISPKSDVWSL GCILYYMTYG KTPFQQIINQ ISKLHAIIDP NHEIEFPDIP EKDLQDVLKC CLKRDPKQRI SIPELLAHPY VQIQTHPVNQ MAKGTTEEMK YVLGQLVGLN SPNSILKAAK TLYEHYSGGE SHNSSSSKTF EKKRGKK
TTK is a serine/threonine kinase closely related to SPK1, PIM 1 and CDC2 serine/threonine kinases. TTK is a dual-specificity kinase, which can also phosphorylate tyrosine. It consists of a common heteromeric core enzyme, which is composed of a catalytic subunit and a constant regulatory subunit, that associates with a variety of regulatory subunits. The constant regulatory subunit A serves as a scaffolding molecule to coordinate the assembly of the catalytic subunit and a variable regulatory B subunit. TTK encodes an alpha isoform of the constant regulatory subunit A. Alternatively spliced transcript variants have been described.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: cancer/testis antigen 96; Dual specificity protein kinase TTK; monopolar spindle 1 kinase; monopolar spindle 1-like 1; OTTHUMP00000016778; OTTHUMP00000220743; phosphotyrosine picked threonine kinase; Phosphotyrosine picked threonine-protein kinase; PYT
Gene Aliases: CT96; ESK; MPH1; MPS1; MPS1L1; PYT; TTK
UniProt ID: (Human) P33981
Entrez Gene ID: (Human) 7272
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support