Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Recommended positive controls: 293T, A431, H1299, HeLaS3, Molt-4, Raji, NIH-3T3.
Predicted reactivity: Mouse (97%), Rat (98%), Zebrafish (85%), Xenopus laevis (91%), Bovine (98%).
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Catalytic subunit of the neuronal tubulin polyglutamylase complex. Modifies alpha- and beta-tubulin, generating side chains of glutamate on the gamma-carboxyl groups of specific glutamate residues within the C-terminal tail of alpha- and beta-tubulin.
⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: bM121M7.1 (novel protein (ortholog of human tubulin tyrosine ligase-like 1 (TTLL1)), isoform 1); catalytic subunit of neural tubulin polyglutamylase; p49; PGs3; Polyglutamylase complex subunit TTLL1; probable tubulin polyglutamylase TTLL1; Tubulin polyglutamylase complex subunit 3; Tubulin polyglutamylase TTLL1; tubulin tyrosine ligase-like family member 1; tubulin tyrosine ligase-like family, member 1; Tubulin--tyrosine ligase-like protein 1; tubulin-tyrosine ligase
Gene Aliases: 6330444E16Rik; AV014541; C22orf7; HS323M22B; TTLL1
UniProt ID: (Human) O95922, (Mouse) Q91V51
Entrez Gene ID: (Human) 25809, (Mouse) 319953
Molecular Function: microtubule or microtubule-binding cytoskeletal protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support