Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
Promotions
View available promotion(s)
Promo Code: P5877545 Don't miss out: buy 3, only pay for 2 Extended to June 20: take advantage of our 3 for 2 promotion. Whether you are restocking essentials or trying something new, now is a great time to get more for your money!. Learn more
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
Immunogen sequence: MELGSCLEG GREAAEEEGE PEVKKRRLLC VEFASVASCD AAVAQCFLAE NDWEMERALN SYFEPPVEES ALERRPETIS EPKTYVDLTN EETTDSTTSK ISPSEDTQQE NGSMFSLITW NIDGLDLNNL SERARGVCSY LALYSPDVIF LQEVIPPYYS YLKKRSSNYE IITGHEEGYF TAIMLKKSRV KLKSQEIIPF PSTKMMRNLL CVHVNVSGNE LCLMTSHLES TRGHAAERMN QLKMVLKKMQ EAPESATVIF AGDTNLRDRE VTRCGGLPNN IVDVWEFLGK PKHCQYTWDT QMNSNLGITA ACKLRFDRIF FRAAAEEGHI IPRSLDLLGL EKLDCGRFPS DHWGLLCNLD IIL (1-362 aa encoded by BC017553)
This gene encodes a member of a superfamily of divalent cation-dependent phosphodiesterases. The encoded protein associates with CD40, tumor necrosis factor (TNF) receptor-75 and TNF receptor associated factors (TRAFs), and inhibits nuclear factor-kappa-B activation. This protein has sequence and structural similarities with APE1 endonuclease, which is involved in both DNA repair and the activation of transcription factors.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 5'-Tyr-DNA phosphodiesterase; 5'-tyrosyl-DNA phosphodiesterase; EAPII; ETS1-associated protein 2; ETS1-associated protein II; MGC111021; MGC9099; RP1-30M3.3; TRAF and TNF receptor associated protein; TRAF and TNF receptor-associated protein; Tyr-DNA phosphodiesterase 2; Tyrosyl-DNA phosphodiesterase 2; Tyrosyl-RNA phosphodiesterase; VPg unlinkase
Gene Aliases: AD-022; AD022; D13Ertd656e; dJ30M3.3; EAP2; EAPII; hTDP2; TDP2; TTRAP
UniProt ID: (Human) O95551, (Mouse) Q9JJX7, (Rat) Q3T1H5
Entrez Gene ID: (Human) 51567, (Mouse) 56196, (Rat) 498749
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support