Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MGLEAATAVG LSDFCSNPDP YVLNLTQEET GLSSDILSYY LLCNRAVSNP FQQRLTLSQR ALANIHSQLL GLEREAVPQF PSAQKPLLSL EETLNVTEGN
TTYH1 is a member of the tweety family of proteins, a family of chloride anion channels containing five transmembrane regions. TTYH1 is a Ca2+-independent, volume-sensitive large conductance chloride (Cl-) channel. TTYH1 is primarily expressed in neural tissue and upregulated in astrocytoma, glioma, and several other cancers. Recent experiments have shown that TTYH1 is an integral endoplasmic reticulum (ER) membrane protein involved in cell proliferation and is thought to play an essential role in embryonic cell growth, possibly through the Ca2+ storage/release process in ER membranes during early development.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: hTTY1; Protein tweety homolog 1; tweety homolog 1
Gene Aliases: TTYH1
UniProt ID: (Human) Q9H313
Entrez Gene ID: (Human) 57348
Molecular Function: ion channel
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support