Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Primary Antibodies
  • Search All
  • Popular Product Areas
  • TaqMan Assays
  • Primary Antibodies
  • Secondary Antibodies
  • Isotype Controls
  • Proteins & Peptides
  • ELISA Kits
  • All Documents & Support
  • Certificates
  • SDS
  • Manuals & Protocols
  • Product FAQs
    Search button Close
            Target of Interest
            Application
            Target Species
            • Order Status
            • Quick Order
            • Sign in
              Sign in
              Don't have an account ? Create Account
              • Account
              • Check Order Status
              • Aspire Member Program
              • Connect: Lab, Data, Apps
              • Custom Products & Projects
              • Services Central

            Disclaimer

            Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

            Loading ...
            • Primary Antibodies ›
            • TUBA1A Antibodies

            Invitrogen

            TUBA1A Polyclonal Antibody

            4 Published Figures
            7 References
            View all (7) TUBA1A antibodies
            Datasheet
            Protocols
            Questions & Answers
            Catalog # PA5-22060
            Web quote
            You can generate a web quote (PDF) by signing in to your account and adding products to your cart. View cart and click the 'Generate a web quote' button. With your web quote document, you can get approval for your order and complete your order online.
             
            Price (USD)
            370.65
            Online Exclusive
            494.00
            Save 123.35 (25%)
            100 µL
             
            Check your price
            Request bulk or custom format
            Join Aspire™ for free antibodies
            Datasheet
            Protocols
            Questions & Answers

            Cite TUBA1A Polyclonal Antibody

            • Antibody Testing Data (10)
            • Published Figures (4)
            Application
            TUBA1A Antibody in Immunocytochemistry (ICC/IF)
            Group 53 Created with Sketch.

            FIGURE: 1 / 14

            TUBA1A Antibody (PA5-22060) in ICC/IF

            Immunofluorescence analysis of alpha Tubulin antibody was performed using 70% confluent log phase LNCaP cells. The cells were fixed with 4% paraformaldehyde for 10 minutes, permeabilized with 0.1% Triton™ X-100 for 10 minutes, and blocked with 1% BSA for 1 hour at room temperature. The cells were labeled with Alpha Tubulin Rabbit Polyclonal Antibody (Product # PA5-22060) at 2 µg/mL in 0.1% BSA and incubated for 3 hours at room temperature and then labeled with Goat anti-Rabbit IgG (Heavy Chain) Superclonal™ Secondary Antibody, Alexa Fluor®... View More
            View Product

            TUBA1A Antibody in Immunocytochemistry (ICC/IF)
            TUBA1A Antibody in Immunocytochemistry (ICC/IF)
            TUBA1A Antibody in Immunocytochemistry (ICC/IF)
            TUBA1A Antibody in Immunohistochemistry (Paraffin) (IHC (P))
            TUBA1A Antibody in Western Blot (WB)
            TUBA1A Antibody in Western Blot (WB)
            TUBA1A Antibody in Western Blot (WB)
            TUBA1A Antibody in Western Blot (WB)
            TUBA1A Antibody in Western Blot (WB)
            TUBA1A Antibody in Western Blot (WB)
            TUBA1A Antibody in Immunocytochemistry (ICC/IF)
            TUBA1A Antibody in Western Blot (WB)
            TUBA1A Antibody in Western Blot (WB)
            TUBA1A Antibody in Western Blot (WB)

            Product Details

            PA5-22060

            Applications
            Tested Dilution
            Publications

            Western Blot (WB)

            1:500-1:20,000
            View 6 publications 6 publications

            Immunohistochemistry (Paraffin) (IHC (P))

            1:100-1:1,000
            -

            Immunocytochemistry (ICC/IF)

            1:100-1:1,000
            View 1 publication 1 publication
            Product Specifications

            Species Reactivity

            Fruit fly, Human, Mouse, Non-human primate, Rat

            Published species

            Human, Mouse

            Host/Isotype

            Rabbit / IgG

            Class

            Polyclonal

            Type

            Antibody

            Immunogen

            Recombinant protein encompassing a sequence within the center region of human alpha Tubulin 1A. The exact sequence is proprietary.

            Conjugate

            Unconjugated

            Form

            Liquid

            Concentration

            1.14 mg/mL

            Purification

            Antigen affinity chromatography

            Storage buffer

            PBS, pH 7, with 20% glycerol

            Contains

            0.025% ProClin 300

            Storage conditions

            Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.

            Shipping conditions

            Ambient (domestic); Wet ice (international)

            RRID

            AB_11154084

            Product Specific Information

            Recommended positive controls: Jurkat, Raji, 293T, A431, HeLa, HepG2, H1299, HCT116, MCF-7, NT2D1, PC-3, U87-MG, NIH-3T3, mouse brain, rat brain, Drosophila.

            Predicted reactivity: Mouse (100%), Rat (100%), Zebrafish (100%), Xenopus laevis (98%), Pig (99%), Rhesus Monkey (100%), Chimpanzee (100%), Bovine (100%).

            Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

            Target Information

            Alpha Tubulin is a part of a heterodimer consisting of alpha and beta tubulin subunits. The alpha and beta tubulins, which are each about 55 kDa MW, are homologous but not identical. Alpha-beta tubulin heterodimer is the basic building block of microtubules, and this intracellular cylindrical filamentous structure is present in almost eukaryotic cells. Microtubules serve as structural supports and lines of transport within the cell, as well as serving a key role in mitosis. Microtubules of the eukaryotic cytoskeleton perform essential and diverse functions and are composed of a heterodimer of alpha and beta tubulins. The alpha and beta tubulins represent the major components of microtubules, while gamma tubulin plays a critical role in the nucleation of microtubule assembly. There are multiple alpha and beta tubulin genes, which are highly conserved among species. The gene that encodes alpha tubulin is highly similar to the mouse and rat Tuba1 gene. Northern blotting studies have shown that the gene expression is predominantly found in morphologically differentiated neurologic cells.

            For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

            Bioinformatics

            Protein Aliases: alpha 1a antibody; alpha 3 antibody; Alpha-tubulin 1; Alpha-tubulin 3; alpha-tubulin 3 antibody; Alpha-tubulin isotype M-alpha-1; B-ALPHA-1 antibody; brain-specific antibody; FLJ25113 antibody; FLJ30169; H2 ALPHA; H2-ALPHA; H2ALPHA; hum-a-tub1; hum-a-tub1 antibody; hum-a-tub2; hum-a-tub2 antibody; LIS3 antibody; M40; MGC117247; MGC16435; OK/SW cl.56; OK/SW-cl.56; OK/SWcl.56; Talpha1 alpha-tubulin; TUBA1A antibody; TUBA3 antibody; TUBB1; TUBB5; Tubulin alpha-1 chain; Tubulin alpha-1A chain; tubulin alpha-1A chain antibody; Tubulin alpha-3 chain; tubulin alpha-3 chain antibody; Tubulin B-alpha-1; tubulin B-alpha-1 antibody; tubulin, alpha 1; tubulin, alpha 1a; tubulin, alpha, brain-specific

            View more View less

            Gene Aliases: B-ALPHA-1; LIS3; Tuba-1; Tuba1; TUBA1A; TUBA3

            View more View less

            UniProt ID: (Human) Q71U36, (Mouse) P68369, (Rat) P68370

            View more View less

            Entrez Gene ID: (Human) 7846, (Mouse) 22142, (Rat) 64158

            View more View less

            Function(s)

            GTPase activity structural molecule activity structural constituent of cytoskeleton protein binding GTP binding protein domain specific binding nucleotide binding protein heterodimerization activity tubulin

            Process(es)

            G2/M transition of mitotic cell cycle cytoskeleton organization microtubule-based process cytoskeleton-dependent intracellular transport cell division protein polymerization metabolic process
            It has to be done as per old AB suggested Products section.
            guarantee_icon

            Performance Guarantee

            If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

            Learn more
            help_icon

            We're here to help

            Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

            Contact tech support
            Ordering Plus Icon Minus Icon
            • Order Status
            • Order Help
            • Quick Order
            • Supply Center
            • eProcurement
            Support Plus Icon Minus Icon
            • Help and Support
            • Contact Us
            • Technical Support Centers
            • Documents and Certificates
            • Report a Site Issue
            Resources Plus Icon Minus Icon
            • Learning Centers
            • Promotions
            • Events and Webinars
            • Social Media
            About Thermo Fisher Plus Icon Minus Icon
            • About Us
            • Careers
            • Investors
            • News
            • Social Responsibility
            • Trademarks
            • Consumer Health Data Privacy Policy
            Our Portfolio Plus Icon Minus Icon
            • Thermo Scientific
            • Applied Biosystems
            • Invitrogen
            • Gibco
            • Ion Torrent
            • Fisher Scientific
            • Unity Lab Services
            • Patheon
            • PPD
            • Terms & Conditions
            • Privacy Information Center
            • Price & Freight Policy
            • Cookie Preferences

              Your choices regarding cookies on this site

              We and our affiliates and vendors use cookies and similar technologies to operate our sites, recognize visitors to our sites, provide secure log-in, collect statistics to optimize site functionality, and deliver content tailored to your interests. Click Accept All to accept all cookies and go directly to the site, click Reject All to reject all but cookies strictly necessary to the functioning of this site (required cookies), or click on Manage Settings to see detailed descriptions of the types of cookies and choose whether to accept certain cookies while on the site.
            © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
            United States flag icon
            United States

            Your items have has been added!


            Host server : magellan-search-green-56756dfb8b-47p2h:80/100.66.77.189:80.
            git-commit: de6905f249d25e22729e613aef254bb544bd8b7c
            git-url: https://github.com/thermofisher/magellan-search
            git-branch: release/2.31.1-2025.06.25-1.0