Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
FIGURE: 1 / 7
MA5-31941 has been tested in indirect ELISA.
TUBB2A belongs to the tubulin family. Tubulin is the major constituent of microtubules. It binds two moles of GTP, one at an exchangeable site on the beta chain and one at a non-exchangeable site on the alpha-chain.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: class IIa beta-tubulin; dJ40E16.7; tubb2a {ECO:0000250|UniProtKB:Q7TMM9}; Tubulin beta class IIa; Tubulin beta-2A chain; tubulin, beta 2A class IIa; tubulin, beta polypeptide; tubulin, beta polypeptide 2
Gene Aliases: CDCBM5; M(beta)2; TUBB; TUBB2; TUBB2A
UniProt ID: (Human) Q13885, (Rat) P85108, (Mouse) Q7TMM9
Entrez Gene ID: (Human) 7280, (Rat) 498736, (Mouse) 22151
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support