Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MNGTRNWCTL VDVHPEDQAA GSVDILRLTL QGELTGDELE HIAQKAGRKT YAMVSSHSAG HSLASELVES HDGHEEIIKV YLKGRSGDKM IHEKNINQLK SEVQYIQEAR NCLQKLREDI SSKLDRNLGD SLHRQEIQVV LEKPNGFSQS PTALYSSPPE VDTCINEDVE SLRKTVQDLL AKLQEAKRQH QSDCVAFEVT LSRYQREAEQ SNVALQREED RVEQKEAEVG ELQRRLLGME TEHQALLAKV REGEVALEEL RSNNADCQAE REKAATLEKE VAGLREKIHH LDDMLKSQQR KVRQMIEQLQ NSKAVIQSKD ATIQELKEKI AYLEAENLEM HDRMEHLIEK QISHGNFSTQ ARAKTENPGS IRISKPPSPK PMPVIRVVET
TUFT1 is involved in the mineralization and structural organization of enamel.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Tuftelin; UNQ865
Gene Aliases: TUFT1
UniProt ID: (Human) Q9NNX1
Entrez Gene ID: (Human) 7286
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support