Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Antibodies Inc
Promotions
View available promotion(s)
Promo Code: P5906365 Don't miss out: buy 3, only pay for 2 Extended to June 27: take advantage of our 3 for 2 promotion. Whether you are restocking essentials or trying something new, now is a great time to get more for your money!. Learn more
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
Promo Code: RACCS25 Save on advanced culture solutions Save up to 22% on optimized formulations to streamline your neural, stem and cancer cell culture. Learn more
For maximum recovery of product, centrifuge the vial prior to removing the cap.
KCNK6 channels play a critical role in maintaining the electrical stability of cells, and their dysfunction can contribute to a variety of pathophysiological conditions. KCNK6, also known as TWIK-2 (Tandem of Pore Domains in a Weak Inwardly Rectifying K+ Channel 2), is a member of the two-pore-domain potassium (K2P) channel family.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: FLJ12282; Potassium channel subfamily K member 6; TWIK-2 two-pore-domain K+ channel
Gene Aliases: D7Ertd764e; Kcnk6; Toss; Twik2
UniProt ID: (Mouse) Q3TBV4
Entrez Gene ID: (Mouse) 52150
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support