Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MAPSGSLAVP LAVMVPLLWG APWTHGRRSN VRVITDENWR ELLEGDWMIE FYAPWCPACQ NLQPEWESFA EWGEDLEVNI AKVDVTEQPG LSGRFIINAL PTIYHCKDGE FRRYQGPRTK KDFINFISDK EWKSIEPVSS WFGPGSVLMS SMSALFQLSM WIRTCHNYFI EDLGLPVWGS YTVFALATLF SGLLLGLCMI FVADCLCPSK RRRPQPYPYP SKKLLSESAQ PLKKVEEEQE ADEEDVSEEE AESKEGTNKD FPQNAIRQRS LGPSLATDKS
TXNDC1 is a thioredoxin (TXN; see MIM 187700)-related protein with disulfide reductase activity (Matsuo et al., 2001 [PubMed 11152479]).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: DKFZP564E1962; protein disulfide isomerase family A, member 11; Protein disulfide-isomerase TMX1; thioredoxin domain containing 1; Thioredoxin domain-containing protein 1; Thioredoxin-related transmembrane protein 1; Transmembrane Trx-related protein
Gene Aliases: PDIA11; PSEC0085; TMX; TMX1; TXNDC; TXNDC1; UNQ235/PRO268
UniProt ID: (Human) Q9H3N1
Entrez Gene ID: (Human) 81542
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support