Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Not known. associated with cell differentiation. The protein encoded by this gene is a member of the thioredoxin family. The exact function of this protein is not known but it is associated with cell differentiation.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ATP binding protein; ATP binding protein associated with cell differentiation; ATP-binding protein associated with cell differentiation; ES cell-related protein; Protein 1-4; Thioredoxin domain-containing protein 9
Gene Aliases: AI098020; APACD; PHLP3; TXNDC9
UniProt ID: (Human) O14530, (Mouse) Q9CQ79
Entrez Gene ID: (Human) 10190, (Rat) 280671, (Mouse) 98258
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support