Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: EILLSADHI IIATGGRPRY PTHIEGALEY GITSDDIFWL KESPGKTLVV GASYVALECA GFLTGIGLDT TIMMRSIPLR GFDQQMSSMV IEHMASHGTR FLRGCAPSRV RRLPDGQLQV TWEDSTTGKE DTGTFDTVLW AIGRVPDTRS LNLEKAGVDT SPDTQKILVD SREATSVPHI YAIGDVVEGR PELTPIAIMA GRLLVQRLFG GSSDLMDYDN VPTTVFTPLE YGCVGLSEEE AVARHGQEHV EVYHAHYKPL EFTVAGRDAS QCYVKMVCLR EPPQLVLGLH FLGPNAGEVT QGFALGIKCG ASYAQVMRTV GIHPTCSEEV VKLRISKRSG LDPTVTGCUG (137-485 aa encoded by BC007489)
The mammalian thioredoxin reductases (TrxRs) are a family of selenocysteine-containing pyridine nucleotide-disulfide oxidoreductases. All the mammalian TrxRs are homologous to glutathione reductase with respect to primary structure including the conserved redox catalytic site (-Cys-Val-Asn-Val-Gly-Cys-) but distinctively with a C-terminal extension containing a catalytically active penultimate selenocysteine (SeCys) residue in the conserved sequence (-Gly-Cys-SeCys-Gly). TrxR is homodimeric protein in which each monomer includes an FAD prosthetic group, a NADPH binding site and a redox catalytic site. Electrons are transferred from NADPH via FAD and the active-site disulfide to C-terminal SeCys-containing redox center, which then reduces the substrate like thioredoxin. The members of TrxR family are 55-58 kilodalton in molecular size and composed of three isoforms including cytosolic TrxR1, mitochondrial TrxR2, and TrxR3, known as Trx and GSSG reductase (TGR). TrxR plays a key role in protection of cells against oxidative stress and redox-regulatory mechanism of transcription factors and various biological phenomena.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Selenoprotein Z; SelZ; Thioredoxin reductase 2, mitochondrial; thioredoxin reductase 3; thioredoxin reductase beta; Thioredoxin reductase TR3; TR beta; TR-beta
Gene Aliases: AA118373; ESTM573010; KIAA1652; SELZ; TGR; TR; TR-BETA; TR3; TRXR2; Trxrd2; TXNRD2
UniProt ID: (Human) Q9NNW7, (Mouse) Q9JLT4, (Rat) Q9Z0J5
Entrez Gene ID: (Human) 10587, (Mouse) 26462, (Rat) 50551
Molecular Function: reductase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support