Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
Immunogen sequence: MPVAGSELP RRPLPPAAQE RDAEPRPPHG ELQYLGQIQH ILRCGVRKDD RTGTGTLSVF GMQARYSLRD EFPLLTTKRV FWKGVLEELL WFIKGSTNAK ELSSKGVKIW DANGSRDFLD SLGFSTREEG DLGPVYGFQW RHFGAEYRDM ESDYSGQGVD QLQRVIDTIK TNPDDRRIIM CAWNPRDLPL MALPPCHALC QFYVVNSELS CQLYQRSGDM GLGVPFNIAS YALLTYMIAH ITGLKPGDFI HTLGDAHIYL NHIEPLKIQL QREPRPFPKL RILRKVEKID DFKAEDFQIE GYNPHPTIKM EMAV (1-313 aa encoded by BC002567)
Thymidylate Synthase (TS) converts deoxyuridine monophosphate (dUMP) to deoxythymidine monophosphate (dTMP), which is essential for DNA biosynthesis. TS is a critical target for the fluoropyrimidines, an important group of antineoplastic drugs that are widely used in the treatment of solid tumors. Both 5-FU and fluorodeoxyuridine are converted in tumor cells to FdUMP which inactivates TS by formation of a ternary covalent complex in the presence of the folate cofactor 5,10-methylenetetrahydrofolate. Expression of TS protein is associated with response to 5-fluorouracil (5-FU) in human colorectal, gastric, head and neck, and breast carcinomas.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: MGC88736; Thymidylate synthase; TS; TSase; tysy
Gene Aliases: HST422; OK/SW-cl.29; TMS; TS; TYMS
UniProt ID: (Human) P04818
Entrez Gene ID: (Human) 7298
Molecular Function: methyltransferase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support