Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Thymidylate Synthase (TS) converts deoxyuridine monophosphate (dUMP) to deoxythymidine monophosphate (dTMP), which is essential for DNA biosynthesis. TS is a critical target for the fluoropyrimidines, an important group of antineoplastic drugs that are widely used in the treatment of solid tumors. Both 5-FU and fluorodeoxyuridine are converted in tumor cells to FdUMP which inactivates TS by formation of a ternary covalent complex in the presence of the folate cofactor 5,10-methylenetetrahydrofolate. Expression of TS protein is associated with response to 5-fluorouracil (5-FU) in human colorectal, gastric, head and neck, and breast carcinomas.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: MGC88736; Thymidylate synthase; TS; TSase; tysy
Gene Aliases: HST422; OK/SW-cl.29; TMS; TS; TYMS
UniProt ID: (Human) P04818
Entrez Gene ID: (Human) 7298
Molecular Function: methyltransferase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support