Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MSAPKLLSLG CIFFPLLLFQ QARAQFPRQC ATVEALRSGM CCPDLSPVSG PGTDRCGSSS GRGRCEAVTA DSRPHSPQYP HDGRDDREVW PLRFFNRTCH CNGNFSGHNC GTCRPGWRGA ACDQRVLIVR RNLLDLSKEE KNHFVRALDM AKRTTHPLFV IATRRSEEIL GPDGNTPQFE NISIYNYFVW THYYSVKKTF LGVGQESFGE VDFSHEGPAF LTWHRYHLLR LEKDMQEMLQ EPSFSLPYWN FATGKNVCDI CTDDLMGSRS NFDSTLISPN SVFSQWRVVC DSLEDYDTLG TLCNSTEDGP IRRNPAGNVA RPMVQRLPEP QDVAQCLEVG LFDTPPFYSN STNSFRNTVE GYSDPTGKYD PAVRSLHNLA HLFLNGTGGQ THLSPNDPIF VLLHTFTDAV FDEWLRRYNA DISTFPLENA PIGHNRQYNM VPFWPPVTNT EMFVTAPDNL GYTYEIQWPS REFSVPEIIA IAVVGALLLV ALIFGTASYL IRARRSMDEA NQPLLTDQYQ CYAEEYEKLQ NPNQSVV
This gene encodes a melanosomal enzyme that belongs to the tyrosinase family and plays an important role in the melanin biosynthetic pathway. Defects in this gene are the cause of rufous oculocutaneous albinism and oculocutaneous albinism type III.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 5,6-dihydroxyindole-2-carboxylic acid oxidase; Catalase B; DHICA oxidase; Glycoprotein 75; Melanoma antigen gp75; MGC120175; MGC149362; RP11-3L8.1; TRP; TRY1; TRY4; TRYP1; Tyrosinase-related protein 1
Gene Aliases: b-PROTEIN; CAS2; CATB; GP75; OCA3; TRP; TRP1; TYRP; TYRP1; TYRRP
UniProt ID: (Human) P17643
Entrez Gene ID: (Human) 7306
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support