Search Thermo Fisher Scientific
Antibody tested in indirect ELISA
Tebuconazole is a systemic fungicide used to control a variety of fungal diseases in crops such as cereals, fruits, and vegetables. It works by inhibiting the production of ergosterol, an essential component of fungal cell membranes. Tebuconazole is effective against a wide range of fungal pathogens, including powdery mildew, rust, and leaf spot. It is also used in non-agricultural settings, such as golf courses and ornamental plantings.